missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-5-fluorophenylacetate, 96%

Product Code. 15409078
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15409078

Brand: Thermo Scientific Alfa Aesar H62561.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 937732-38-2 | C9H8BrFO2 | 247.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937732-38-2
Molecular Formula C9H8BrFO2
Molecular Weight (g/mol) 247.063
InChI Key AJSCFDHWZUZCNE-UHFFFAOYSA-N
Synonym methyl 2-3-bromo-5-fluorophenyl acetate, methyl 3-bromo-5-fluorophenyl acetate
PubChem CID 59290393
IUPAC Name methyl 2-(3-bromo-5-fluorophenyl)acetate
SMILES COC(=O)CC1=CC(=CC(=C1)Br)F

Specifications

Quantity 250 mg
Formula Weight 247.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 3-bromo-5-fluorophenylacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.