missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-tetralone, 96%, Thermo Scientific Chemicals

Product Code. 15409077
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15409077 1 g 1g
15419077 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409077 Supplier Thermo Scientific Alfa Aesar Supplier No. H52381.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 66361-67-9 | Molecular Formula: C10H9BrO | Molecular Weight: 225.09 g/mol

6-bromo-1-tetralone used in Wittig-Horner reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-bromo-1-tetralone used in Wittig-Horner reaction.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66361-67-9
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.09
MDL Number MFCD04114378
InChI Key OSDHOOBPMBLALZ-UHFFFAOYSA-N
Synonym 6-bromo-1-tetralone, 6-bromo-3,4-dihydronaphthalen-1 2h-one, 6-bromotetral-1-one, 6-bromo-tetral-1-on, 6-bromotetralone, 1 2h-naphthalenone, 6-bromo-3,4-dihydro, 6-bromo-1,2,3,4-tetrahydronaphthalen-1-one, 6-bromo-3,4-dihydro-1 2h-naphthalenone, 6-bromoteralone, 6-bromo-tetralone
PubChem CID 10105069
IUPAC Name 6-bromo-3,4-dihydro-2H-naphthalen-1-one
SMILES BrC1=CC2=C(C=C1)C(=O)CCC2

Specifications

Melting Point 40°C to 42°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 225.09
Percent Purity 96%
Chemical Name or Material 6-Bromo-1-tetralone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.