missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-indanone, 97%

Product Code. 15460438
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460438 5 g 5g
15450438 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15460438 Supplier Thermo Scientific Alfa Aesar Supplier No. H54003.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14548-39-1 | C9H7BrO | 211.058 g/mol

6-Bromo-1-indanone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1-indanone is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14548-39-1
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD02179286
InChI Key SEQHEDQNODAFIU-UHFFFAOYSA-N
Synonym 6-bromo-1-indanone, 6-bromoindanone, 6-bromo-2,3-dihydro-1h-inden-1-one, 6-bromo-indan-1-one, 6-bromoindan-1-one, 1-indanone, 6-bromo, 1h-inden-1-one, 6-bromo-2,3-dihydro, 6-bromo indanone, 6-bromo-indanone
PubChem CID 139778
IUPAC Name 6-bromo-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C=CC(=C2)Br

Specifications

Melting Point 110°C
Quantity 5 g
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 6-Bromo-1-indanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.