missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-[1,8]naphthyridin-2(1H)-one, Thermo Scientific™

Product Code. 10507970
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10507970 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10507970 Supplier Thermo Scientific Maybridge Supplier No. MAY00219DA
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 72754-05-3
Packaging Amber Glass Bottle
Molecular Formula C8H5BrN2O
MDL Number MFCD07781151
Quantity 1 g
Synonym 6-bromo-1,8-naphthyridin-2-ol, 6-bromo-1,8-naphthyridin-2 1h-one, 6-bromo-1,8 naphthyridin-2 1h-one, 6-bromo-1,8 naphthyridin-2-ol, 6-bromo-2-hydroxy-1,8-naphthyridine, 1,8-naphthyridin-2 1h-one, 6-bromo, acmc-1bihh, ksc496s1t, 6-bromopyridino 2,3-b pyridin-2-ol, 6-bromo-1,8naphthyridin-2 1h-one
InChI Key BCEWGGGNOQNYKK-UHFFFAOYSA-N
SMILES BrC1=CN=C2NC(=O)C=CC2=C1
IUPAC Name 6-bromo-1H-1,8-naphthyridin-2-one
Molecular Weight (g/mol) 225.05
PubChem CID 12520144
Formula Weight 225.04
Percent Purity 95%
Chemical Name or Material 6-Bromo-[1, 8]naphthyridin-2(1H)-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.