missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,8-naphthyridin-2(1H)-one, 98%

Product Code. 11414430
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414430 250 mg 250mg
11424430 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414430 Supplier Thermo Scientific Alfa Aesar Supplier No. H50042.MD
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 72754-05-3 | C8H5BrN2O | 225.05 g/mol

6-Bromo-1,8-naphthyridin-2(1H)-one is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1,8-naphthyridin-2(1H)-one is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72754-05-3
Molecular Formula C8H5BrN2O
Molecular Weight (g/mol) 225.05
MDL Number MFCD07781151
InChI Key BCEWGGGNOQNYKK-UHFFFAOYSA-N
PubChem CID 12520144
IUPAC Name 6-bromo-1H-1,8-naphthyridin-2-one
SMILES BrC1=CN=C2NC(=O)C=CC2=C1

Specifications

Melting Point >300°C
Quantity 250 mg
Synonym 6-bromo-1,8-naphthyridin-2-ol, 6-bromo-1,8-naphthyridin-2 1h-one, 6-bromo-1,8 naphthyridin-2 1h-one, 6-bromo-1,8 naphthyridin-2-ol, 6-bromo-2-hydroxy-1,8-naphthyridine, 1,8-naphthyridin-2 1h-one, 6-bromo, acmc-1bihh, ksc496s1t, 6-bromopyridino 2,3-b pyridin-2-ol, 6-bromo-1,8naphthyridin-2 1h-one
Solubility Information Insoluble in water.
Formula Weight 225.05
Percent Purity 98%
Chemical Name or Material 6-Bromo-1,8-naphthyridin-2(1H)-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.