missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific™

Product Code. p-7030573
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 15467765

Brand: Thermo Scientific Alfa Aesar H30436.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene is used in Synthesis and evaluation of synthetic retinoid derivatives as inducers of stem cell differentiation.

Solubility
Insoluble in water

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27452-17-1
Molecular Formula C14H19Br
Molecular Weight (g/mol) 267.21
MDL Number MFCD05664407
InChI Key NLOOVMVNNNYLFS-UHFFFAOYSA-N
Synonym 6-bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, naphthalene, 6-bromo-1,2,3,4-tetrahydro-1,1,4,4-tetramethyl, 1,1,4,4-tetramethyl-6-bromotetralin, 6-bromo-1,1,4,4-tetramethyl-tetralin, 6-bromo-1,1,4,4 tetramethy 1,2,3,4-tetrahydronaphthalene, 6-bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphtalene, 2-bromo-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene, 2-bromo-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaph-thalene, 2-bromo-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphthalene
PubChem CID 226685
IUPAC Name 6-bromo-1,1,4,4-tetramethyl-2,3-dihydronaphthalene
SMILES CC1(CCC(C2=C1C=CC(=C2)Br)(C)C)C

Specifications

Melting Point 34°C to 37°C
Quantity 10 g
Solubility Information Insoluble in water
Formula Weight 267.21
Percent Purity 97%
Chemical Name or Material 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.