missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 6-Bnz-cAMP sodium salt

Product Code. 18765708
Change view
Click to view available options
:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18765708 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18765708 Supplier Tocris Bioscience™ Supplier No. 5255/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell permeable cAMP analog

6-Bnz-cAMP sodium salt is a cell permeable cAMP analog; selectively activates cAMP-dependent PKA but not Epac signaling pathways. Acts synergistically with 8-CPT-2Me-cAMP (Cat. No. 1645) to inhibit vascular smooth muscle cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1135306-29-4
Quantity 1 mg
Product Type 6-Bnz-cAMP sodium salt
Purity 98%
Molecular Weight (g/mol) M.W. (Observed): 455.29
Inhibitors 6-Bnz-cAMP sodium salt

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.