missing translation for 'onlineSavingsMsg'
Learn More

6-Biopterin, MedChemExpress

Product Code. 16496417
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496417 5 mg 5mg
16452826 25 mg 25mg
16414766 100 mg 100mg
16403688 10 mg 10mg
16400077 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496417 Supplier Medchem Express Supplier No. HY102015
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 6-Biopterin (L-Biopterin), a pterin derivative, is a NO synthase cofactor.

6-Biopterin (L-Biopterin), a pterin derivative, is a NO synthase cofactor.

Specifications

Chemical Name or Material 6-Biopterin
CAS 22150-76-1
Color White
Purity Grade Notes Research
Molecular Formula C9H11N5O3
Quantity 5 mg
Synonym L-Biopterin
Solubility Information DMSO : 12.5 mg/mL (52.69 mM; ultrasonic and warming and heat to 75°C)
SMILES O=C1N=C(N)NC2=NC=C([C@@H](O)[C@@H](O)C)N=C12
Molecular Weight (g/mol) 237.22
Formula Weight 237.22
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.