missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyloxy Warfarin, TRC

Product Code. 30342298
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342298 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342298 Supplier Toronto Research Chemicals Supplier No. TRCB2885605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Benzyloxy Warfarin
CAS 30992-68-8
Molecular Formula C26 H22 O5
Quantity 5 mg
InChI Formula InChI=1S/C26H22O5/c1-17(27)14-21(19-10-6-3-7-11-19)24-25(28)22-15-20(12-13-23(22)31-26(24)29)30-16-18-8-4-2-5-9-18/h2-13,15,21,28H,14,16H2,1H3
SMILES CC(=O)CC(C1=C(O)c2cc(OCc3ccccc3)ccc2OC1=O)c4ccccc4
IUPAC Name 4-hydroxy-3-(3-oxo-1-phenylbutyl)-6-phenylmethoxychromen-2-one
Molecular Weight (g/mol) 414.45
Formula Weight 414.1467
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.