missing translation for 'onlineSavingsMsg'
Learn More

6-(Benzyloxy)-2-naphthylboronic acid, 97%, Thermo Scientific™

Product Code. 10304682
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10304682 1 g Amber Glass Bottle 1g
10394672 250 mg Amber Glass Bottle 250mg
10206004 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10304682 Supplier Thermo Scientific Maybridge Supplier No. MO00847DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 152915-83-8
Molecular Formula C17H15BO3
Molecular Weight (g/mol) 278.114
MDL Number MFCD04115402
InChI Key ZVHAZMUBLCEMGG-UHFFFAOYSA-N
Synonym 6-benzyloxy-2-naphthylboronic acid, 6-benzyloxynaphthalene-2-boronic acid, 6-benzyloxy naphthalen-2-ylboronic acid, boronic acid,b-6-phenylmethoxy-2-naphthalenyl, 6-benzyloxy-2-naphthaleneboronic acid, 6-benzyloxy naphthalen-2-yl boronic acid, 6-phenylmethoxynaphthalen-2-yl boronic acid, acmc-209d8c, 6-benzyloxy-2-naphthylboronicacid
PubChem CID 2794628
IUPAC Name (6-phenylmethoxynaphthalen-2-yl)boronic acid
SMILES B(C1=CC2=C(C=C1)C=C(C=C2)OCC3=CC=CC=C3)(O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 278.11
Percent Purity 97%
Chemical Name or Material 6-(Benzyloxy)-2-naphthylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.