missing translation for 'onlineSavingsMsg'
Learn More

6-Benzylaminopurine, MedChemExpress

Product Code. 16488526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488526 Supplier Medchem Express Supplier No. HYB0941
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 6-Benzylaminopurine is a cytokinin that elicits plant growth and development, also is an inhibitor of respiratory kinase in plants, increases post-harvest life of green vegetables.

6-Benzylaminopurine is a cytokinin that elicits plant growth and development, also is an inhibitor of respiratory kinase in plants, increases post-harvest life of green vegetables.

Specifications

Chemical Name or Material 6-Benzylaminopurine
Physical Form Solid
CAS 1214-39-7
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H11N5
Synonym Benzyladenine 6-BAP N6-Benzyladenine
Solubility Information DMSO : 50 mg/mL (221.98 mM; Need ultrasonic)
SMILES C12=NC=NC(NCC3=CC=CC=C3)=C1NC=N2
Molecular Weight (g/mol) 225.25
Formula Weight 225.25
Percent Purity 98.13%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.