missing translation for 'onlineSavingsMsg'
Learn More

6-(Benzylamino)pyridine-3-boronic acid pinacol ester, 95%

Product Code. 15417695
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417695 250 mg 250mg
11377329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417695 Supplier Thermo Scientific Alfa Aesar Supplier No. H27463.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1073354-27-4 | C18H23BN2O2 | 310.204 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073354-27-4
Molecular Formula C18H23BN2O2
Molecular Weight (g/mol) 310.204
MDL Number MFCD06798270
InChI Key BWIHFPRFKBKWMU-UHFFFAOYSA-N
Synonym 6-benzylamino pyridine-3-boronic acid pinacol ester, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, n-benzyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, amtb753, 6-benzylamino pyridine-3-boronicacidpinacolester, 2-benzylamino pyridine-5-boronic acid pinacol ester, n-phenylmethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-pyridinamine, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-amine, 2-pyridinamine, n-phenylmethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 44755164
IUPAC Name N-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)NCC3=CC=CC=C3

Specifications

Melting Point 135°C to 138°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 310.2
Percent Purity 95%
Chemical Name or Material 6-(Benzylamino)pyridine-3-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.