missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyladenine, 99%, Thermo Scientific Chemicals

Catalog Number 11417666
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1214-39-7
C12H11N5
225.26
MFCD00005572
NWBJYWHLCVSVIJ-UHFFFAOYSA-N
6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
62389
CHEBI:29022
N-benzyl-7H-purin-6-amine
C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1
This item is not returnable. View return policy
1214-39-7
C12H11N5
225.26
MFCD00005572
NWBJYWHLCVSVIJ-UHFFFAOYSA-N
6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
62389
CHEBI:29022
N-benzyl-7H-purin-6-amine
C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1

Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

Solubility
Soluble in water, methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.

Notes
Incompatible with strong oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 229°C to 233°C
Assay Percent Range 99%
Quantity 1 g
Beilstein 19406
Solubility Information Soluble in water,methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.
Formula Weight 225.26
Percent Purity 99%
Chemical Name or Material 6-Benzyladenine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'