missing translation for 'onlineSavingsMsg'
Learn More

6-Benzoylhexanoic acid, 94%, Thermo Scientific Chemicals

Product Code. 11328165
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328165 1 g 1g
11338165 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328165 Supplier Thermo Scientific Alfa Aesar Supplier No. L08431.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7472-43-7 | Molecular Formula: C13H16O3 | Molecular Weight: 220.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7472-43-7
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.27
MDL Number MFCD00039608
InChI Key DOQWHEUDAHLEPT-UHFFFAOYSA-N
Synonym 6-benzoylhexanoic acid, 7-oxo-7-phenyl-heptanoic acid, 6-benzoyl hexanoic acid, benzeneheptanoic acid, z-oxo, 7-oxo-7-phenylheptanoicacid, #, benzeneheptanoic acid,, ae-oxo, alpha-oxo benzeneheptanoic acid, benzeneheptanoic acid,alpha-oxo
PubChem CID 344830
IUPAC Name 7-oxo-7-phenylheptanoic acid
SMILES OC(=O)CCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 83°C to 86°C
Quantity 1 g
Beilstein 2694641
Formula Weight 220.27
Percent Purity 94%
Chemical Name or Material 6-Benzoylhexanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.