missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Azauridine, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 6-Azauridine |
| CAS | 54-25-1 |
| Molecular Formula | C8 H11 N3 O6 |
| Quantity | 100 mg |
| Synonym | 2-β-D-ribofuranosyl-1,2,4-triazine-3,5(2H,4H)dione, 6-Azauracil 1-riboside, 6-AzUr, Riboazauracil |
| InChI Formula | InChI=1S/C8H11N3O6/c12-2-3-5(14)6(15)7(17-3)11-8(16)10-4(13)1-9-11/h1,3,5-7,12,14-15H,2H2,(H,10,13,16)/t3-,5-,6-,7-/m1/s1 |
| SMILES | OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2N=CC(=O)NC2=O |
| IUPAC Name | 2-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]-1,2,4-triazine-3,5-dione |
| Molecular Weight (g/mol) | 245.19 |
| Formula Weight | 245.0848 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction