missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 6-Azauracil, 99%

Product Code. 10021740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021740 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10021740 Supplier Thermo Scientific Chemicals Supplier No. 153970050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 461-89-2 | C3H3N3O2 | 113.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 461-89-2
CAS Min % 98.5
CAS Max % 100.0
Melting Point 276°C to 280°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C3H3N3O2
MDL Number MFCD00006456
Quantity 5 g
Beilstein 26, V,5, 304
Synonym 6-azauracil, 1,2,4-triazine-3,5 2h,4h-dione, azauracil, 1,2,4-triazine-3,5-diol, 4 6-azauracil, as-triazine-3,5-diol, as-triazine-3,5 2h,4h-dione, 1,2,4-triazine-3,5-dione, usaf cb-30, unii-i14twn70lr
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,dmso
InChI Key SSPYSWLZOPCOLO-UHFFFAOYSA-N
SMILES O=C1NN=CC(=O)N1
IUPAC Name 2H-1,2,4-triazine-3,5-dione
Molecular Weight (g/mol) 113.08
PubChem CID 68037
ChEBI CHEBI:53745
Formula Weight 113.08
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 6-Azauracil, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 6-Azauracil
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.