missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoquinolyl-N-hydroxysuccinimidyl Carbamate (90%), TRC

Product Code. 30138244
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138244 250 mg 250mg
17991805 100 mg 100mg
17904365 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138244 Supplier Toronto Research Chemicals Supplier No. TRCA629140250MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148757-94-2
Molecular Formula C14 H11 N3 O4
Molecular Weight (g/mol) 285.25
Synonym N-6-Quinolinylcarbamic Acid 2,5-Dioxo-1-pyrrolidinyl Ester, 1-[[(6-Quinolinylamino)carbonyl]oxy]-2,5-pyrrolidinedione, AHC Reagent, AccQ.Fluor, AccQTag;
IUPAC Name (2,5-dioxopyrrolidin-1-yl) N-quinolin-6-ylcarbamate
SMILES O=C(Nc1ccc2ncccc2c1)ON3C(=O)CCC3=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H11N3O4/c18-12-5-6-13(19)17(12)21-14(20)16-10-3-4-11-9(8-10)2-1-7-15-11/h1-4,7-8H,5-6H2,(H,16,20)
Formula Weight 285.075
Chemical Name or Material 6-Aminoquinolyl-N-hydroxysuccinimidyl Carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.