missing translation for 'onlineSavingsMsg'
Learn More

6-Aminopyridine-2-carboxylic acid, 95%

Product Code. 11345265
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345265 1 g 1g
11335265 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345265 Supplier Thermo Scientific Alfa Aesar Supplier No. H31861.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 23628-31-1 | C6H6N2O2 | 138.13 g/mol

It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

Solubility
Soluble in hot water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23628-31-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00233711
InChI Key NMCKJFCJIHCHIS-UHFFFAOYSA-N
Synonym 6-aminopicolinic acid, 2-aminopyridine-6-carboxylic acid, 6-amino-2-pyridinecarboxylic acid, 6-amino-2-picolinic acid, 6-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-amino, 2-amino-6-pyridinecarboxylic acid, picolinic acid, 6-amino, pubchem7686, acmc-209g4h
PubChem CID 351652
IUPAC Name 6-aminopyridine-2-carboxylic acid
SMILES NC1=CC=CC(=N1)C(O)=O

Specifications

Melting Point ∽295°C (decomposition)
Odor Odorless
Quantity 1 g
Solubility Information Soluble in hot water.
Formula Weight 138.13
Percent Purity 95%
Chemical Name or Material 6-Aminopyridine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.