missing translation for 'onlineSavingsMsg'
Learn More

(+)-6-Aminopenicillanic Acid, TRC

Product Code. 30322750
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322750 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322750 Supplier Toronto Research Chemicals Supplier No. TRCA61889510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Aminopenicillanic Acid
CAS 551-16-6
Purity Grade Notes HPLC
Molecular Formula C8 H12 N2 O3 S
Quantity 10 g
Synonym 6-Aminopenicillanic Acid, (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid, Amoxicillin Trihydrate Imp. A (EP), Ampicillin Sodium Imp. A (EP), Benzylpenicillin Potassium Imp. A (EP), Cloxacillin Sodium Imp. C (EP), Dicloxacillin Sodium Imp. C (EP), Flucloxacillin Sodium Imp. C (EP), Oxacillin Sodium Imp. A (EP), Phenoxymethylpenicillin Potassium Imp. C (EP), Piperacillin Sodium Imp. H (EP)
InChI Formula InChI=1S/C8H12N2O3S/c1-8(2)4(7(12)13)10-5(11)3(9)6(10)14-8/h3-4,6H,9H2,1-2H3,(H,12,13)/t3-,4+,6-/m1/s1
SMILES CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(=O)O
IUPAC Name (2S,5R,6R)-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular Weight (g/mol) 216.26
Formula Weight 216.0569
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.