missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-6-Aminopenicillanic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Aminopenicillanic Acid |
| CAS | 551-16-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 H12 N2 O3 S |
| Quantity | 1 g |
| Synonym | 6-Aminopenicillanic Acid, (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid, Amoxicillin Trihydrate Imp. A (EP), Ampicillin Sodium Imp. A (EP), Benzylpenicillin Potassium Imp. A (EP), Cloxacillin Sodium Imp. C (EP), Dicloxacillin Sodium Imp. C (EP), Flucloxacillin Sodium Imp. C (EP), Oxacillin Sodium Imp. A (EP), Phenoxymethylpenicillin Potassium Imp. C (EP), Piperacillin Sodium Imp. H (EP) |
| InChI Formula | InChI=1S/C8H12N2O3S/c1-8(2)4(7(12)13)10-5(11)3(9)6(10)14-8/h3-4,6H,9H2,1-2H3,(H,12,13)/t3-,4+,6-/m1/s1 |
| SMILES | CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(=O)O |
| IUPAC Name | (2S,5R,6R)-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
| Molecular Weight (g/mol) | 216.26 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?