missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 6-Aminopenicillanic acid, 98%

Product Code. p-7020932
Click to view available options
Quantity:
10g
50g
250g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11430004

Brand: Thermo Scientific™ A13056.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
6-Aminopenicillanic acid is the basic unit in penicillin’s, which is obtained from the fermentation brew of the Penicillium mold. It acts as a starting material for the preparation of many semisynthetic penicillins. Further, it serves as apharmaceutical intermediate. In addition to this, it is used in antibiotics research and experimental application.

Solubility
Soluble in water and hydrochoric acid.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

Purity 98%
CAS 551-16-6
Content And Storage Keep cold
Molecular Weight (g/mol) 216.26
Product Type 6-Aminopenicillanic acid
Quantity 10g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.