missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 6-Aminopenicillanic acid, 98%
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
6-Aminopenicillanic acid is the basic unit in penicillin’s, which is obtained from the fermentation brew of the Penicillium mold. It acts as a starting material for the preparation of many semisynthetic penicillins. Further, it serves as apharmaceutical intermediate. In addition to this, it is used in antibiotics research and experimental application.
Solubility
Soluble in water and hydrochoric acid.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
6-Aminopenicillanic acid is the basic unit in penicillin’s, which is obtained from the fermentation brew of the Penicillium mold. It acts as a starting material for the preparation of many semisynthetic penicillins. Further, it serves as apharmaceutical intermediate. In addition to this, it is used in antibiotics research and experimental application.
Solubility
Soluble in water and hydrochoric acid.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Purity | 98% |
| CAS | 551-16-6 |
| Content And Storage | Keep cold |
| Molecular Weight (g/mol) | 216.26 |
| Product Type | 6-Aminopenicillanic acid |
| Quantity | 250g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?