missing translation for 'onlineSavingsMsg'
Learn More

6-Aminopenicillanic acid, 96%

Product Code. 10456900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10456900 100 g Glass bottle 100g
10568863 10 g Glass bottle 10g
10234560 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10456900 Supplier Thermo Scientific Acros Supplier No. 104251000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 551-16-6 | C8H12N2O3S | 216.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 551-16-6
Molecular Formula C8H12N2O3S
Molecular Weight (g/mol) 216.26
MDL Number MFCD00005176
InChI Key NGHVIOIJCVXTGV-ALEPSDHESA-N
Synonym 6-aminopenicillanic acid, 6-apa, penicin, penin, aminopenicillanic acid, 6-aps, phenacyl 6-aminopenicillinate, 6-aminopenicillamine acid, 6-aminopenicillanate, 2s,5r,6r-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid
PubChem CID 11082
ChEBI CHEBI:16705
IUPAC Name (2S,5R,6R)-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 197°C to 203°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 454
Specific Rotation + 276.3
Formula Weight 216.25
Percent Purity 96%
Physical Form Fine Powder
Chemical Name or Material 6-Aminopenicillanic acid, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Aminopenicillanic acid
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.