missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinic acid, 98%

Product Code. 10616594 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10616594 1 g Glass bottle 1g
10236450 5 g Glass bottle 5g
10523761 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10616594 Supplier Thermo Scientific Acros Supplier No. 202540010
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3167-49-5 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3167-49-5
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00006326
InChI Key ZCIFWRHIEBXBOY-UHFFFAOYSA-N
Synonym 6-aminonicotinic acid, 3-pyridinecarboxylic acid, 6-amino, 2-amino-5-pyridinecarboxylic acid, 6-amino-nicotinic acid, 6-amino nicotinic acid, 2-aminopyridine-5-carboxylic acid, nicotinic acid, 6-amino, unii-9vr2o1ok60, 6-amino-3-pyridinecarboxylic acid, 6-aminopyridine 3-carboxylic acid
PubChem CID 18496
ChEBI CHEBI:68583
IUPAC Name 6-aminopyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2N(C5H3N)COOH
Quantity 1 g
Beilstein 22, 542
Merck Index 15, 452
Solubility Information Solubility in water: ca 1.0g/L (20°C). Other solubilities: sparingly soluble in most solvents,5.0g/L (20°C) in ethanol
Formula Weight 138.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-Aminonicotinic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.