missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinamide, MP Biomedicals™

Product Code. 11416492 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416492 1 g 1g
11426492 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416492 Supplier MP Biomedicals Supplier No. 0215010301
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 329-89-5
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006327
InChI Key ZLWYEPMDOUQDBW-UHFFFAOYSA-N
Synonym 6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid
PubChem CID 9500
ChEBI CHEBI:74514
IUPAC Name 6-aminopyridine-3-carboxamide
SMILES NC(=O)C1=CC=C(N)N=C1

Specifications

Quantity 1 g
Formula Weight 137.1
Physical Form Solid
Chemical Name or Material 6-Aminonicotinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.