missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoindole, 97%, Thermo Scientific Chemicals

Catalog Number 10775442
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5318-27-4
C8H8N2
132.17
MFCD02093959
MIMYTSWNVBMNRH-UHFFFAOYSA-N
6-aminoindole, 6-amino-1h-indole, 6-amino indole, 1h-indol-6-ylamine, indole-6-ylamine, 1h-indol-6-amin, 6-indolamine, indol-6-amine, pubchem7311
256096
1H-indol-6-amine
NC1=CC=C2C=CNC2=C1
This item is not returnable. View return policy
5318-27-4
C8H8N2
132.17
MFCD02093959
MIMYTSWNVBMNRH-UHFFFAOYSA-N
6-aminoindole, 6-amino-1h-indole, 6-amino indole, 1h-indol-6-ylamine, indole-6-ylamine, 1h-indol-6-amin, 6-indolamine, indol-6-amine, pubchem7311
256096
1H-indol-6-amine
NC1=CC=C2C=CNC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 66.0°C to 68.0°C
Color White to Brown
Boiling Point 161.0°C to 166.0°C (2.3 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 132.17
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminoindole
Show More Show Less
compliance-icons
Product Identifier
  • 6-Aminoindole, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'