missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Aminoindan-1-one, 97%, Thermo Scientific™
Click to view available options
5g; Glass bottle
Specifications
CAS | 69975-65-1 |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Molecular Formula | C9H9NO |
MDL Number | MFCD00100747 |
Quantity | 5g |
Synonym | 6-aminoindanone, 6-aminoindan-1-one, 6-amino-1-indanone, 6-amino-2,3-dihydro-1h-inden-1-one, 6-amino-indan-1-one, 1h-inden-1-one, 6-amino-2,3-dihydro, 6-amino indanone, ksc497g1b, 6-azanyl-2,3-dihydroinden-1-one |
Show More |
Safety and Handling
Product Identifier
- 6-Aminoindan-1-one
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'