missing translation for 'onlineSavingsMsg'
Learn More

6-(Aminocarbonyl)-3-cyclohexene-1-carboxylic Acid, TRC

Product Code. 30136133
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.50g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136133 2.5 g 2.50g
30134681 1 g 1g
30010936 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136133 Supplier Toronto Research Chemicals Supplier No. TRCA3577652.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2028-12-8
Molecular Formula C8 H11 N O3
Molecular Weight (g/mol) 169.178
Synonym 3-Cyclohexene-1-carboxylic acid, 6-carbamoyl- (6CI,7CI,8CI), 6-(Aminocarbonyl)-3-cyclohexene-1-carboxylic acid, 6-Carbamoylcyclohex-3-enecarboxylic acid, NSC 191910
IUPAC Name 6-carbamoylcyclohex-3-ene-1-carboxylic acid
SMILES NC(=O)C1CC=CCC1C(=O)O

Specifications

Analyte or Component Names 1,2,3,6-Tetrahydrophthalamic acid
Shipping Condition Room Temperature
InChI Formula InChI=1S/C8H11NO3/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-2,5-6H,3-4H2,(H2,9,10)(H,11,12)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 2.5 g
Formula Weight 169.0739
Chemical Name or Material 1,2,3,6-Tetrahydrophthalamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.