missing translation for 'onlineSavingsMsg'
Learn More

6-Aminohexanoic acid, 99%

Product Code. 11417746
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417746 100 g 100g
11427746 500 g 500g
11437746 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417746 Supplier Thermo Scientific Alfa Aesar Supplier No. A14719.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

6-aminohexanoic acid, CAS # 60-32-2, is a synthetic lysine derivative with inhibitory activity against fibrinolysis. It shows hematologic activity. | CAS: 60-32-2 | C6H13NO2 | 131.175 g/mol

6-Aminohexanoic acid is used as an intermediate in the polymerization of Nylon-6. It is involved in the extraction of aldehydes from the reaction mixture as well as to improve the solubilization of membrane proteins in electrophoresis. It acts as an inhibitor of fibrinolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 6-aminohexanoic acid, an epsilon-amino acid, shows an antifibrinolytic and hematologic activity
  • This compound competitively inhibits the activation of plasminogen; it consequently reduces the conversion of plasminogen to plasmin (fibrinolysin), an enzyme responsible for the degradation of fibrin clots, fibrinogen, and other plasma proteins, including the procoagulant factors V and VIII

Application

  • It has been used as an intermediate in the polymerization of Nylon-6
  • It is used for biomolecular applications to extract aldehydes from the reaction mixture and to increase membrane protein solubilization in electrophoresis experiments
  • It has been described to inhibit plasminogen binding to activated platelets
  • This compound can bind and inactivate carboxypeptidase B
  • This compound can also inhibit the activation of the first component of the complement system
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-32-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00008238
InChI Key SLXKOJJOQWFEFD-UHFFFAOYSA-N
Synonym 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
PubChem CID 564
ChEBI CHEBI:16586
IUPAC Name 6-aminohexanoic acid
SMILES C(CCC(=O)O)CCN

Specifications

Melting Point 210°C to 212°C
Assay Percent Range ≥99%
Linear Formula H2N(CH2)5CO2H
Quantity 100 g
Beilstein 906872
Merck Index 14,432
Solubility Information Soluble in water. Slightly soluble in alcohol. Practically insoluble in ether and chloroform.
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material 6-Aminocaproic Acid
compliance-icons
Product Identifier
  • 6-Aminohexanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 6-Aminohexanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.