missing translation for 'onlineSavingsMsg'
Learn More

6-Aminobenzo[b]thiophene-1,1-dioxide, Thermo Scientific™

Product Code. 10465512
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 20503-40-6
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD00173800
InChI Key KRUCRVZSHWOMHC-UHFFFAOYSA-N
Synonym 6-aminobenzo b thiophene 1,1-dioxide, 6-amino-1h-1lambda-6-benzo b thiophene-1,1-dione, 6-amino-1,1-dioxobenzo b thiophene, benzo b thiophen-6-amine,1,1-dioxide, 6-amino-1lambda6-benzothiophene-1,1-dione, 6-amino-1??-benzothiophene-1,1-dione, 6-amino-1, pubchem14478, maybridge1_005192, 6-aminobenzo b thiole-1,1-dione
PubChem CID 2779855
IUPAC Name 1,1-dioxo-1-benzothiophen-6-amine
SMILES C1=CC(=CC2=C1C=CS2(=O)=O)N

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 181.21
Percent Purity Tech.
Grade Technical
Chemical Name or Material 6-Aminobenzo[b]thiophene 1, 1-dioxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'