missing translation for 'onlineSavingsMsg'
Learn More

6-Aminobenzo[b]thiophene-1,1-dioxide, Thermo Scientific™

Product Code. 10069271
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10069271 1 g Amber Glass Bottle 1g
10465512 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10069271 Supplier Thermo Scientific Maybridge Supplier No. MWP00044DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 20503-40-6
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD00173800
InChI Key KRUCRVZSHWOMHC-UHFFFAOYSA-N
Synonym 6-aminobenzo b thiophene 1,1-dioxide, 6-amino-1h-1lambda-6-benzo b thiophene-1,1-dione, 6-amino-1,1-dioxobenzo b thiophene, benzo b thiophen-6-amine,1,1-dioxide, 6-amino-1lambda6-benzothiophene-1,1-dione, 6-amino-1??-benzothiophene-1,1-dione, 6-amino-1, pubchem14478, maybridge1_005192, 6-aminobenzo b thiole-1,1-dione
PubChem CID 2779855
IUPAC Name 1,1-dioxo-1-benzothiophen-6-amine
SMILES C1=CC(=CC2=C1C=CS2(=O)=O)N

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 181.21
Percent Purity Tech.
Grade Technical
Chemical Name or Material 6-Aminobenzo[b]thiophene 1, 1-dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.