missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-5-nitrouracil, TRC

Product Code. 30322717
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322717 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322717 Supplier Toronto Research Chemicals Supplier No. TRCA618495500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Amino-5-nitrouracil
CAS 3346-22-3
Molecular Formula C4 H4 N4 O4
Quantity 500 mg
InChI Formula InChI=1S/C4H4N4O4/c5-2-1(8(11)12)3(9)7-4(10)6-2/h(H4,5,6,7,9,10)
SMILES NC1=C(C(=O)NC(=O)N1)[N+](=O)[O-]
IUPAC Name 6-amino-5-nitro-1H-pyrimidine-2,4-dione
Molecular Weight (g/mol) 172.1
Formula Weight 172.0233
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.