missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-5-bromoquinoline, 97%

Product Code. 15499678
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499678 1 g 1g
15419688 250 mg 250mg
15409688 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499678 Supplier Thermo Scientific Alfa Aesar Supplier No. H63150.03
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 50358-42-4 | C9H7BrN2 | 223.073 g/mol

6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

Solubility
Slightly soluble in water. Soluble in alcohol, ether, and carbon disulfide.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 50358-42-4
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.073
MDL Number MFCD00234503
InChI Key MODLGTLYXJGDCH-UHFFFAOYSA-N
PubChem CID 12741232
IUPAC Name 5-bromoquinolin-6-amine
SMILES C1=CC2=C(C=CC(=C2Br)N)N=C1

Spezifikation

Color Yellow to Brown
Quantity 1 g
Synonym 6-amino-5-bromoquinoline, 6-quinolinamine, 5-bromo, 5-bromo-6-quinolinamine, 6-quinolinamine,5-bromo, 5-bromo-quinolin-6-ylamine, 6-quinolinamine, 5-bromo-9ci
Solubility Information Slightly soluble in water. Soluble in alcohol,ether,and carbon disulfide.
Formula Weight 223.07
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Amino-5-bromoquinoline

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.