missing translation for 'onlineSavingsMsg'
Learn More

(6-Amino-3-pyridinyl)methanol, 97%, Thermo Scientific™

Product Code. 10196692
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10196692 1 g 1g
10094962 250 mg 250mg
10603835 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10196692 Supplier Thermo Scientific Maybridge Supplier No. MO07396DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 113293-71-3
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
InChI Key TXPRFSOGPYITOT-UHFFFAOYSA-N
PubChem CID 2794829
IUPAC Name (6-aminopyridin-3-yl)methanol
SMILES C1=CC(=NC=C1CO)N

Specifications

Assay Percent Range 97%
Quantity 1 g
Synonym 6-aminopyridin-3-yl methanol, 6-amino-3-pyridinyl methanol, 6-amino-3-pyridinemethanol, 2-amino-5-hydroxymethylpyridine, 6-amino-pyridin-3-yl-methanol, 2-amino-5-hydroxymethyl pyridine, 2-amino-5-pyridinemethanol, 3-pyridinemethanol, 6-amino, 3-pyridinemethanol,6-amino, 6-amino-3-hydroxymethylpyridine
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material (6-Amino-3-pyridinyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.