missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-thiouracil Hydrate, TRC

Product Code. 30331547
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331547 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331547 Supplier Toronto Research Chemicals Supplier No. TRCA63045525G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Amino-2-thiouracil Hydrate
CAS 65802-56-4
Molecular Formula C4 H5 N3 O S . H2 O
Quantity 25 g
InChI Formula InChI=1S/C4H5N3OS.H2O/c5-2-1-3(8)7-4(9)6-2;/h1H,(H4,5,6,7,8,9);1H2
SMILES O.NC1=CC(=O)NC(=S)N1
IUPAC Name 6-amino-2-sulfanylidene-1H-pyrimidin-4-one;hydrate
Molecular Weight (g/mol) 161.18
Formula Weight 161.0259
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.