missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-methyl-2H-indazole, 97%, Thermo Scientific™

Product Code. 10441420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10441420 1 g Glass bottle 1g
10637040 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10441420 Supplier Thermo Scientific Acros Supplier No. 432430010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 50593-30-1
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD00464866
InChI Key MHCWLERQNFATHZ-UHFFFAOYSA-N
Synonym 2-methyl-2h-indazol-6-amine, 6-amino-2-methyl-2h-indazole, 2-methyl-2h-indazol-6-ylamine, 2-methyl-2h-indazole-6-amine, 2h-indazol-6-amine, 2-methyl, 2-methyl-2h-indazole-6-ylamine, 2-methyl-6-indazolamine, 6-amino-2-methyl-indazole, 2-methylindazol-6-yl amine, 2h-indazol-6-amine,2-methyl
PubChem CID 590220
IUPAC Name 2-methylindazol-6-amine
SMILES CN1C=C2C=CC(=CC2=N1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 162°C to 163°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 147.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2-methyl-2H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 6-Amino-2-methyl-2H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.