missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole, 97%, Thermo Scientific™

Product Code. 10423068 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10423068 1 g 1g
10649209 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423068 Supplier Thermo Scientific Acros Supplier No. 431120010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1053655-59-6
Molecular Formula C12H15N3O
Molecular Weight (g/mol) 217.27
MDL Number MFCD10699171
InChI Key BVMGAUOJVUEFSV-UHFFFAOYSA-N
Synonym 1-tetrahydro-2h-pyran-2-yl-1h-indazol-6-amine, 6-amino-1-tetrahydro-2h-pyran-2-yl-1h-indazole, 1-tetrahydro-pyran-2-yl-1h-indazol-6-ylamine, 1-oxan-2-yl indazol-6-amine, 1-oxan-2-yl-1h-indazol-6-amine, 6-amino-1-tetrahydropyranyl-1h-indazole
PubChem CID 44118233
IUPAC Name 1-(oxan-2-yl)indazol-6-amine
SMILES C1CCOC(C1)N2C3=C(C=CC(=C3)N)C=N2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 92°C to 95°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 217.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 6-Amino-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.