missing translation for 'onlineSavingsMsg'
Learn More

6-Alpha-Methyl Prednisolone, TRC

Product Code. 30137136
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137136 50 mg 50mg
30008067 1 g 1g
18072250 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137136 Supplier Toronto Research Chemicals Supplier No. TRCM32593450MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-43-2
Molecular Formula C22 H30 O5
Molecular Weight (g/mol) 374.47
Synonym Methylprednisolone, Methylprednisolone Hydrogen Succinate Imp. A (EP), Methylprednisolone Acetate Imp. B (EP), Pregna-1,4-diene-3,20-dione, 11,17,21-trihydroxy-6-methyl-, (6α,11β)-, Pregna-1,4-diene-3,20-dione, 11β,17,21-trihydroxy-6α-methyl- (7CI,8CI), (6α,11β)-11,17,21-Trihydroxy-6-methylpregna-1,4-diene-3,20-dione, 11β,17,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione, 11β,17α,21-Trihydroxy-6α-methyl-1,4-pregnadiene-3,20-dione, 6α-Methyl-11β,17α,21β-trihydroxy-1,4-pregnadiene-3,20-dione, 6α-Methylprednisolone, A-Methapred, Besonia, Dopomedrol, Duralone, M-Prednisol, Medesone, Medorol, Medralone, Medrate, Medrol, Medrone, Metastab, Methypred, Metilbetasone, Metilprednisolone, Metipred, Metrisone, Moderin, NSC 19987, Noretona, Prednol, Prednol L, Promacortine, SW 196788, Solimedrol, Suprametil, U 7532, Urbason, Urbasone, Urbazone, Wyacort, δ1-6α-Methylhydrocortisone
IUPAC Name (6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)CO)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H30O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,17,19,23,25,27H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,17-,19+,20-,21-,22-/m0/s1
Formula Weight 374.2093
Percent Purity >95%
Chemical Name or Material Methylprednisolone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.