missing translation for 'onlineSavingsMsg'
Learn More

6-Acetylbenzodioxane, TRC

Product Code. 30381253
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381253 50 mg
30383407 10 mg
30386150 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381253 Supplier Toronto Research Chemicals Supplier No. TRCD45490350MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2879-20-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.18
Synonym 1,4-Benzodioxin, Ethanone Deriv., 1-(2,3-Dihydro-1,4-benzodioxin-6-yl)ethanone, 1-(2,3-Dihydro-1,4-benzodioxin-6-yl)ethan-1-one, 1-(2,3-Dihydrobenzo[1,4]dioxin-6-yl)ethanone, 1-(Benzodioxan-6-yl)ethanone, 3,4-Ethylenedioxyacetophenone, 3',4'-Ethylenedioxyacetophenone, 6-Acetyl-1,4-benzodioxan, 6-Acetyl-1,4-benzodioxane, 6-Acetyl-2,3-dihydro-1,4-benzodioxin
SMILES CC(=O)c1ccc2OCCOc2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H10O3/c1-7(11)8-2-3-9-10(6-8)13-5-4-12-9/h2-3,6H,4-5H2,1H3
Formula Weight 178.063
Chemical Name or Material 6-Acetylbenzodioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.