missing translation for 'onlineSavingsMsg'
Learn More

6-Acetyl-1,4-benzodioxane, 98%, Thermo Scientific™

Product Code. 11432907
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432907 100 g 100g
11412907 5 g 5g
11422907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432907 Supplier Thermo Scientific Alfa Aesar Supplier No. A15506.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2879-20-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00006823
InChI Key HGVWMTAIIYNQSI-UHFFFAOYSA-N
PubChem CID 76143
IUPAC Name 1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
SMILES CC(=O)C1=CC=C2OCCOC2=C1

Specifications

Melting Point 81°C to 84°C
Quantity 100 g
Beilstein 164125
Synonym 6-acetyl-1,4-benzodioxane, 1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 1,4-benzodioxan-6-yl methyl ketone, 1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 6-acetyl-1,4-benzodioxan, ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl, ethanone,1-2,3-dihydro-1,4-benzodioxin-6-yl, 1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethan-1-one, 6-acetyl-2h,3h-benzo e 1,4-dioxin
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material 6-Acetyl-1,4-benzodioxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.