missing translation for 'onlineSavingsMsg'
Learn More

6,9-Diamino-2-ethoxyacridine lactate monohydrate, 95%, Thermo Scientific Chemicals

Product Code. 10493745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10493745 100 g Glass bottle 100g
10026023 5 g Glass bottle 5g
10750672 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10493745 Supplier Thermo Scientific Acros Supplier No. 406381000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6402-23-9 , 1837-57-6
Molecular Formula C3H6O3·H2O
Molecular Weight (g/mol) 361.39
MDL Number MFCD00149646
InChI Key NYEPHMYJRNWPLA-UHFFFAOYSA-N
Synonym ethacridine lactate monohydrate, ethodin, ethacridine lactate hydrate, ethacridine lactate salt, dsstox_cid_26274, dsstox_rid_81501, dsstox_gsid_46274, 7-ethoxyacridine-3,9-diamine; 2-hydroxypropanoic acid; hydrate, rivanol monohydrate
PubChem CID 165457
IUPAC Name 7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid;hydrate
SMILES CCOC1=CC2=C(C3=C(C=C(C=C3)N)N=C2C=C1)N.CC(C(=O)O)O.O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 243°C to 245°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 3771
Solubility Information Solubility in water: .. Other solubilities: slowly soluble in 15 parts water,soluble in 9 parts boiling water,soluble in 110 parts alcohol (22°C),soluble in 100 parts alcohol (20°C)
Formula Weight 361.39
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 6, 9-Diamino-2-ethoxyacridine lactate monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • 6,9-Diamino-2-ethoxyacridine lactate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.