missing translation for 'onlineSavingsMsg'
Learn More

6,8-Difluoro-1,4-dihydro-1-(N-methylformamido)-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester, TRC

Product Code. 30384146
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384146 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384146 Supplier Toronto Research Chemicals Supplier No. TRCD44685525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6,8-Difluoro-1,4-dihydro-1-(N-methylformamido)-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester
CAS 158585-86-5
Molecular Formula C19H22F2N4O4
Quantity 25 g
InChI Formula InChI=1S/C19H22F2N4O4/c1-4-29-19(28)13-10-25(23(3)11-26)16-12(18(13)27)9-14(20)17(15(16)21)24-7-5-22(2)6-8-24/h9-11H,4-8H2,1-3H3
SMILES CCOC(=O)C1=CN(N(C)C=O)c2c(F)c(N3CCN(C)CC3)c(F)cc2C1=O
IUPAC Name ethyl 6,8-difluoro-1-[formyl(methyl)amino]-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylate
Molecular Weight (g/mol) 408.4
Formula Weight 408.1609
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.