missing translation for 'onlineSavingsMsg'
Learn More

6,8-Dichlorooctanoic Acid Ethyl Ester, TRC

Product Code. 30385131
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385131 100 g
30372944 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385131 Supplier Toronto Research Chemicals Supplier No. TRCD435555100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-64-0
Molecular Formula C10 H18 Cl2 O2
Molecular Weight (g/mol) 241.15
Synonym Ethyl 6,8-Dichlorooctanoate
SMILES CCOC(=O)CCCCC(Cl)CCCl

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H18Cl2O2/c1-2-14-10(13)6-4-3-5-9(12)7-8-11/h9H,2-8H2,1H3
Chemical Name or Material 6,8-Dichlorooctanoic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.