missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethylpterin, TRC

Product Code. 30383613
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383613 10 g
30378056 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383613 Supplier Toronto Research Chemicals Supplier No. TRCD47791510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-55-2
Molecular Formula C8 H9 N5 O
Molecular Weight (g/mol) 191.19
Synonym 2-Amino-6,7-dimethyl-4(3H)-pteridinone, 2-Amino-6,7-dimethyl-4(1H)-pteridinone, 2-Amino-6,7-dimethyl-4-hydroxypteridine, 2-Amino-4-hydroxy-6,7-dimethylpteridine
IUPAC Name 2-amino-6,7-dimethylpteridin-4-ol
SMILES Cc1nc2nc(N)nc(O)c2nc1C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H9N5O/c1-3-4(2)11-6-5(10-3)7(14)13-8(9)12-6/h1-2H3,(H3,9,11,12,13,14)
Formula Weight 191.0807
Chemical Name or Material 6,7-Dimethylpterin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.