missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxyquinazoline-2,4-dione, TRC

Product Code. 30380874
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380874 1 g -
30377450 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380874 Supplier Toronto Research Chemicals Supplier No. TRCD4749501G
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28888-44-0
Molecular Formula C10 H10 N2 O4
Molecular Weight (g/mol) 222.2
Synonym 6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione, Doxazosin Mesilate Imp. D (EP), 2,4(1H,3H)-Quinazolinedione, 6,7-dimethoxy-, 6,7-Dimethoxy-2,4(1H,3H)-quinazolinedione, 2,5-Dihydroxy-6,7-dimethoxyquinazoline, 6,7-Dimethoxy-1H-quinazoline-2,4-dione, 6,7-Dimethoxyquinazoline-2,4-diol, 6,7-Dimethoxyquinazoline-2,4-dione
IUPAC Name 6,7-dimethoxy-1H-quinazoline-2,4-dione
SMILES COc1cc2NC(=O)NC(=O)c2cc1OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H10N2O4/c1-15-7-3-5-6(4-8(7)16-2)11-10(14)12-9(5)13/h3-4H,1-2H3,(H2,11,12,13,14)
Formula Weight 222.0641
Chemical Name or Material 6,7-dimethoxyquinazoline-2,4(1H,3H)-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.