missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride hydrate, 99+%, Thermo Scientific™

Catalog Number p-4510364
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20232-39-7
C11H14ClNO2
227.69
MFCD11099370
PQXVEYYRJHMTEV-UHFFFAOYSA-N
6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, chloride, 6,7-dimethoxy-3,4-dihydro-isoquinoline hydrochloride, pubchem17358, ksc496s5h, 3,4-dihydro-6,7-dimethoxyisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hydro-chloride, 6,7-dimethoxy-3,4-dihydroisoquinolinehydrochloridehydrate
2724664
Cl.COC1=CC2=C(C=NCC2)C=C1OC
This item is not returnable. View return policy
20232-39-7
C11H14ClNO2
227.69
MFCD11099370
PQXVEYYRJHMTEV-UHFFFAOYSA-N
6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, chloride, 6,7-dimethoxy-3,4-dihydro-isoquinoline hydrochloride, pubchem17358, ksc496s5h, 3,4-dihydro-6,7-dimethoxyisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hydro-chloride, 6,7-dimethoxy-3,4-dihydroisoquinolinehydrochloridehydrate
2724664
Cl.COC1=CC2=C(C=NCC2)C=C1OC
Melting Point 198.0°C to 202.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
IUPAC Name 6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride
Formula Weight 227.69
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material 6, 7-Dimethoxy-3, 4-dihydroisoquinoline hydrochloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'