missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-2,4-quinazolinediamine, TRC

Product Code. 30376814
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30376814 25 mg 25mg
30376501 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30376814 Supplier Toronto Research Chemicals Supplier No. TRCD44660025MG
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60547-96-8
Molecular Formula C10 H12 N4 O2
Molecular Weight (g/mol) 220.23
Synonym 2,4-Diamino-6,7-dimethoxyquinazoline
IUPAC Name 6,7-dimethoxyquinazoline-2,4-diamine
SMILES COc1cc2nc(N)nc(N)c2cc1OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H12N4O2/c1-15-7-3-5-6(4-8(7)16-2)13-10(12)14-9(5)11/h3-4H,1-2H3,(H4,11,12,13,14)
Formula Weight 220.096
Chemical Name or Material 6,7-Dimethoxy-2,4-quinazolinediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.