missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate, 98%, Thermo Scientific™

Product Code. 11445710
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11445710

Brand: Thermo Scientific Alfa Aesar H25941.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4784-02-5

Specifications

Melting Point 352°C to 354°C
Molecular Formula C10H10N2O4
MDL Number MFCD09953682
Quantity 1 g
Synonym 6,7-dimethoxyquinoxaline-2,3 1h,4h-dione, 2,3-dihydroxy-6,7-dimethoxyquinoxaline, 6,7-dimethoxyquinoxaline-2,3-diol, chembl42430, 6,7-dimethoxy-1,4-dihydro-2,3-quinoxalinedione, acmc-20aogl, 2,3-quinoxalinedione,1,4-dihydro-6,7-dimethoxy, 6,7-dimethoxy-1,4-dihydro-quinoxaline-2,3-dione, 2,3-quinoxalinedione, 1,4-dihydro-6,7-dimethoxy, 6,7-dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate
InChI Key AVEIMNFUPXGCJQ-UHFFFAOYSA-N
SMILES COC1=C(C=C2C(=C1)NC(=O)C(=O)N2)OC
IUPAC Name 6,7-dimethoxy-1,4-dihydroquinoxaline-2,3-dione
Molecular Weight (g/mol) 222.20
PubChem CID 613780
Formula Weight 240.22 (222.20 Anhydrous)
Percent Purity 98%
Chemical Name or Material 6,7-Dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.