missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydroxyflavone, TRC

Product Code. 30385489
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385489 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385489 Supplier Toronto Research Chemicals Supplier No. TRCD6790080.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 38183-04-9
Molecular Formula C15 H10 O4
Quantity 0.5 mg
InChI Formula InChI=1S/C15H10O4/c16-11-7-14(9-4-2-1-3-5-9)19-15-8-13(18)12(17)6-10(11)15/h1-8,17-18H
SMILES Oc1cc2OC(=CC(=O)c2cc1O)c3ccccc3
IUPAC Name 6,7-dihydroxy-2-phenyl-chromen-4-one
Molecular Weight (g/mol) 254.24
Formula Weight 254.0579
Chemical Name or Material 6,7-Dihydroxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.