missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydroxy-4-methylcoumarin, 97%

Product Code. 11463874
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463874 25 g 25g
11453874 5 g 5g
11473874 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463874 Supplier Thermo Scientific Alfa Aesar Supplier No. A11902.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 529-84-0 | C10H8O4 | 192.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-84-0
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00006859
InChI Key KVOJTUXGYQVLAJ-UHFFFAOYSA-N
Synonym 4-methylesculetin, 6,7-dihydroxy-4-methylcoumarin, methylesculetin, 6,7-dihydroxy-4-methyl-2h-chromen-2-one, 4-methylaesculetin, 4-methylesculetol, 4-methyl-6,7-dihydroxycoumarin, 2h-1-benzopyran-2-one, 6,7-dihydroxy-4-methyl, coumarin, 6,7-dihydroxy-4-methyl, 6,7-dihydroxy-4-methyl-2h-benzopyran-2-one
PubChem CID 5319502
IUPAC Name 6,7-dihydroxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=CC(=C(C=C12)O)O

Specifications

Melting Point ∼275°C (decomposition)
Quantity 25 g
Beilstein 162550
Formula Weight 192.17
Percent Purity 97%
Chemical Name or Material 6,7-Dihydroxy-4-methylcoumarin

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.